CAS RN 27643-15-8
IUPAC Name:
1,2,3-thiadiazole-4-carbaldehyde
Synonyms:
1,2,3-thiadiazole-4-carbaldehyde
Molecular Weight (g/mol):
114.12
Molecular Formula:
C3H2N2OS
InChi Key:
JNEBZFFTOLBIKJ-UHFFFAOYSA-N
SMILES:
O=CC1=CSN=N1
1
–
1
of
1
results
1
1,2,3-Thiadiazole-4-carbaldehyde, Thermo Scientific™
CAS: 27643-15-8 Molecular Formula: C3H2N2OS Molecular Weight (g/mol): 114.122 InChI Key: JNEBZFFTOLBIKJ-UHFFFAOYSA-N Synonym: 1,2,3-thiadiazole-4-carbaldehyde,1,2,3-thiadiazole-4-carboxaldehyde,4-thiadiazolecarboxaldehyde,acmc-1cb1x,1,2,3-thiadiazol-4-carboxaldehyde PubChem CID: 2737271 IUPAC Name: thiadiazole-4-carbaldehyde SMILES: C1=C(N=NS1)C=O