missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Propanetricarbonitrile 98.0+%, TCI America™

Catalog No. p-7135183
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16301G 1 g
P16305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P16301G Supplier TCI America Supplier No. P16301G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 62872-44-0
Molecular Formula C6H5N3
Molecular Weight (g/mol) 119.13
MDL Number MFCD09038541
InChI Key MNAMONWYCZEPTE-UHFFFAOYSA-N
Synonym 3-Cyanopentanedinitrile
PubChem CID 21248644
IUPAC Name propane-1,2,3-tricarbonitrile
SMILES N#CCC(CC#N)C#N

Specifications

Melting Point 51°C
Color White-Yellow
Boiling Point 158°C
Quantity 1 g
UN Number 3439
Formula Weight 119.13
Percent Purity ≥98.0% (GC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3-Propanetricarbonitrile