accessibility menu, dialog, popup

UserName

NNN

CAS RN 62872-44-0

IUPAC Name: propane-1,2,3-tricarbonitrile
Synonyms: propane-1,2,3-tricarbonitrile
Molecular Weight (g/mol): 119.13
Molecular Formula: C6H5N3
InChi Key: MNAMONWYCZEPTE-UHFFFAOYSA-N
SMILES: N#CCC(CC#N)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)