missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3-Propanetricarbonitrile 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 51°C |
| Color | White-Yellow |
| Boiling Point | 158°C |
| Quantity | 1 g |
| UN Number | 3439 |
| Formula Weight | 119.13 |
| Percent Purity | ≥98.0% (GC) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,2,3-Propanetricarbonitrile |
Safety and Handling
| TSCA | No |