missing translation for 'onlineSavingsMsg'
Learn More

1-(2,3-Epoxypropyl)-2-nitroimidazole, 97%

Catalog No. AAH66536MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66536MD 250 mg
AAH6653603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66536MD Supplier Thermo Scientific Chemicals Supplier No. H66536MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13551-90-1 | C6H7N3O3 | 169.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13551-90-1
Molecular Formula C6H7N3O3
Molecular Weight (g/mol) 169.14
MDL Number MFCD20527194
InChI Key SYFMSLVOHQZYEB-UHFFFAOYSA-N
Synonym 1-2,3-epoxypropyl-2-nitroimidazole, 2-nitro-1-oxiran-2-ylmethyl imidazole, 2-nitro-1-oxiran-2-ylmethyl-1h-imidazole, 1-glycidyl-2-nitro-1h-imidazole, 1-oxiranylmethyl-2-nitroimidazole, 2-nitro-1-oxiranylmethyl-1h-imidazole, 1-oxiranylmethyl-2-nitro-1h-imidazole
PubChem CID 334973
IUPAC Name 2-nitro-1-(oxiran-2-ylmethyl)imidazole
SMILES C1C(O1)CN2C=CN=C2[N+](=O)[O-]

Specifications

Quantity 250 mg
Beilstein 612095
Formula Weight 169.14
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(2,3-Epoxypropyl)-2-nitroimidazole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only