CAS RN 13551-90-1
IUPAC Name:
2-nitro-1-[(oxiran-2-yl)methyl]-1H-imidazole
Synonyms:
2-nitro-1-(oxiran-2-ylmethyl)imidazole
Molecular Weight (g/mol):
169.14
Molecular Formula:
C6H7N3O3
InChi Key:
SYFMSLVOHQZYEB-UHFFFAOYNA-N
SMILES:
[O-][N+](=O)C1=NC=CN1CC1CO1
1
–
1
of
1
results
1
Promotions Available
1-(2,3-Epoxypropyl)-2-nitroimidazole, 97%
1-(2,3-Epoxypropyl)-2-nitroimidazole, 97%
CAS: 13551-90-1 Molecular Formula: C6H7N3O3 Molecular Weight (g/mol): 169.14 MDL Number: MFCD20527194 InChI Key: SYFMSLVOHQZYEB-UHFFFAOYSA-N Synonym: 1-2,3-epoxypropyl-2-nitroimidazole,2-nitro-1-oxiran-2-ylmethyl imidazole,2-nitro-1-oxiran-2-ylmethyl-1h-imidazole,1-glycidyl-2-nitro-1h-imidazole,1-oxiranylmethyl-2-nitroimidazole,2-nitro-1-oxiranylmethyl-1h-imidazole,1-oxiranylmethyl-2-nitro-1h-imidazole PubChem CID: 334973 IUPAC Name: 2-nitro-1-(oxiran-2-ylmethyl)imidazole SMILES: C1C(O1)CN2C=CN=C2[N+](=O)[O-]