missing translation for 'onlineSavingsMsg'
Learn More

1-(2,3-Dichlorophenyl)piperazine monohydrochloride, 98+%, Thermo Scientific™

Catalog No. AAL1869706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1869706 5 g
AAL1869714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1869706 Supplier Thermo Scientific Chemicals Supplier No. L1869706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
This product is intended for forensic and research purposes and it is an aripiprazole intermediate.

Solubility
Soluble in water, DMSO and methanol.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Chemical Identifiers

CAS 119532-26-2
Molecular Formula C10H13Cl3N2
Molecular Weight (g/mol) 267.578
MDL Number MFCD00190238
InChI Key CYQFNNSFAGXCEC-UHFFFAOYSA-N
Synonym 1-2,3-dichlorophenyl piperazine hydrochloride, 1-2,3-dichlorophenyl piperazine monohydrochloride, 1-2,3-dichlorophenyl-piperazine hydrochloride, 2,3-dichlorophenylpiperazine hydrochloride, 1-2,3-dichlorophenyl-piperazine monohydrochloride, 1-2,3-dichlorophenyl piperazinehydrochloride, piperazine, 1-2,3-dichlorophenyl-, monohydrochloride, 1-2,3-dichlorophenyl piperazine;hydrochloride, 2,3-dichlorophenyl piperazine, chloride, zlchem 392
PubChem CID 11630372
IUPAC Name 1-(2,3-dichlorophenyl)piperazine;hydrochloride
SMILES C1CN(CCN1)C2=C(C(=CC=C2)Cl)Cl.Cl

Specifications

Melting Point 248°C to 250°C
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in water,DMSO and methanol.
Formula Weight 267.59
Percent Purity ≥98%
Chemical Name or Material 1-(2,3-Dichlorophenyl)piperazine monohydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only