accessibility menu, dialog, popup

UserName

CAS RN 119532-26-2

IUPAC Name: 1-(2,3-dichlorophenyl)piperazine hydrochloride
Synonyms: 2,3-dichlorophenylpiperazine hydrochloride
Molecular Weight (g/mol): 267.58
Molecular Formula: C10H13Cl3N2
InChi Key: CYQFNNSFAGXCEC-UHFFFAOYSA-N
SMILES: Cl.ClC1=CC=CC(N2CCNCC2)=C1Cl
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)