missing translation for 'onlineSavingsMsg'
Learn More

1-(2,3-Dichlorophenyl)piperazine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7127849
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
Catalog No. Quantity
D366025G 25 g
1 options

Catalog No. D366025G

Supplier: TCI America D366025G

Only null left

Chemical Identifiers

CAS 119532-26-2
Molecular Formula C10H13Cl3N2
Molecular Weight (g/mol) 267.578
MDL Number MFCD00190238
InChI Key CYQFNNSFAGXCEC-UHFFFAOYSA-N
Synonym 1-2,3-dichlorophenyl piperazine hydrochloride, 1-2,3-dichlorophenyl piperazine monohydrochloride, 1-2,3-dichlorophenyl-piperazine hydrochloride, 2,3-dichlorophenylpiperazine hydrochloride, 1-2,3-dichlorophenyl-piperazine monohydrochloride, 1-2,3-dichlorophenyl piperazinehydrochloride, piperazine, 1-2,3-dichlorophenyl-, monohydrochloride, 1-2,3-dichlorophenyl piperazine;hydrochloride, 2,3-dichlorophenyl piperazine, chloride, zlchem 392
PubChem CID 11630372
IUPAC Name 1-(2,3-dichlorophenyl)piperazine;hydrochloride
SMILES C1CN(CCN1)C2=C(C(=CC=C2)Cl)Cl.Cl

Specifications

Melting Point 247°C
Color White-Yellow
Quantity 25 g
Formula Weight 267.58
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2,3-Dichlorophenyl)piperazine Hydrochloride
EINECSNumber (5)-6790
TSCA No