missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3-Butanetriol 90.0+%, TCI America™
$766.13 - $3,057.05
Chemical Identifiers
| CAS | 4435-50-1 |
|---|---|
| Molecular Formula | C4H10O3 |
| Molecular Weight (g/mol) | 106.121 |
| MDL Number | MFCD00059662 |
| InChI Key | YAXKTBLXMTYWDQ-UHFFFAOYSA-N |
| Synonym | 1,2,3-Trihydroxybutane |
| PubChem CID | 20497 |
| IUPAC Name | butane-1,2,3-triol |
| SMILES | CC(C(CO)O)O |
Chemical Identifiers
| 4435-50-1 | |
| 106.121 | |
| YAXKTBLXMTYWDQ-UHFFFAOYSA-N | |
| 20497 | |
| CC(C(CO)O)O |
| C4H10O3 | |
| MFCD00059662 | |
| 1,2,3-Trihydroxybutane | |
| butane-1,2,3-triol |
Specifications
| 4435-50-1 | |
| 175°C | |
| MFCD00059662 | |
| 1,2,3-Trihydroxybutane | |
| CC(C(CO)O)O | |
| 106.121 | |
| 106.12 | |
| Liquid |
| Yellow | |
| C4H10O3 | |
| 5 mL | |
| YAXKTBLXMTYWDQ-UHFFFAOYSA-N | |
| butane-1,2,3-triol | |
| 20497 | |
| ≥90.0% (GC) | |
| 1,2,3-Butanetriol |
Safety and Handling
TSCA : No