missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3-Butanetriol 90.0+%, TCI America™
Supplier: TCI America B091825ML
Specifications
| 1,2,3-Butanetriol | |
| Yellow | |
| C4H10O3 | |
| 25 mL | |
| YAXKTBLXMTYWDQ-UHFFFAOYSA-N | |
| butane-1,2,3-triol | |
| 20497 | |
| ≥90.0% (GC) |
| 4435-50-1 | |
| 175°C | |
| MFCD00059662 | |
| 1,2,3-Trihydroxybutane | |
| CC(C(CO)O)O | |
| 106.121 | |
| 106.12 | |
| Liquid |
Chemical Identifiers
| 4435-50-1 | |
| 106.121 | |
| YAXKTBLXMTYWDQ-UHFFFAOYSA-N | |
| 20497 | |
| CC(C(CO)O)O |
| C4H10O3 | |
| MFCD00059662 | |
| 1,2,3-Trihydroxybutane | |
| butane-1,2,3-triol |
Safety and Handling
TSCA : No