missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 1,2,3,5-Tetra-O-acetyl-beta-L-ribofuranose, 98%

CAS: 144490-03-9 | C13H18O9 | 318.278 g/mol

$76.26 - $651.40

Chemical Identifiers

CAS 144490-03-9
Molecular Formula C13H18O9
Molecular Weight (g/mol) 318.278
MDL Number MFCD00674547
InChI Key IHNHAHWGVLXCCI-UMSGYPCISA-N
Synonym 1,2,3,5-tetra-o-acetyl-beta-l-ribofuranose, 1,2,3,5-tetra-o-acetyl-b-l-ribofuranose, 2s,3s,4s,5r-3,4,5-tris acetyloxy oxolan-2-yl methyl acetate, beta-l-ribofuranose 1,2,3,5-tetra-o-acetate, beta-l-ribofuranose tetraacetate, 1,2,3,5-tetra-o-acetyl-?-l-ribofuranose, 2s,3s,4s,5r-3,4,5-triacetyloxyoxolan-2-yl methyl acetate, 2r,3s,4s,5s-5-acetoxymethyl tetrahydrofuran-2,3,4-triyl triacetate
PubChem CID 69431757
IUPAC Name [(2S,3R,4R,5S)-3,4,5-triacetyloxyoxolan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6607403
View Documents
Thermo Scientific Chemicals
J6607403
1 g
Each for $76.26
Only null left
 
AAJ6607406
View Documents
Thermo Scientific Chemicals
J6607406
5 g
Each for $363.66
Only null left
 
AAJ6607409
View Documents
Thermo Scientific Chemicals
J6607409
10 g
Each for $651.40
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

144490-03-9
318.278
IHNHAHWGVLXCCI-UMSGYPCISA-N
69431757
CC(=O)OCC1C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C
C13H18O9
MFCD00674547
1,2,3,5-tetra-o-acetyl-beta-l-ribofuranose, 1,2,3,5-tetra-o-acetyl-b-l-ribofuranose, 2s,3s,4s,5r-3,4,5-tris acetyloxy oxolan-2-yl methyl acetate, beta-l-ribofuranose 1,2,3,5-tetra-o-acetate, beta-l-ribofuranose tetraacetate, 1,2,3,5-tetra-o-acetyl-?-l-ribofuranose, 2s,3s,4s,5r-3,4,5-triacetyloxyoxolan-2-yl methyl acetate, 2r,3s,4s,5s-5-acetoxymethyl tetrahydrofuran-2,3,4-triyl triacetate
[(2S,3R,4R,5S)-3,4,5-triacetyloxyoxolan-2-yl]methyl acetate

Specifications

144490-03-9
White
MFCD00674547
1,2,3,5-tetra-o-acetyl-beta-l-ribofuranose, 1,2,3,5-tetra-o-acetyl-b-l-ribofuranose, 2s,3s,4s,5r-3,4,5-tris acetyloxy oxolan-2-yl methyl acetate, beta-l-ribofuranose 1,2,3,5-tetra-o-acetate, beta-l-ribofuranose tetraacetate, 1,2,3,5-tetra-o-acetyl-?-l-ribofuranose, 2s,3s,4s,5r-3,4,5-triacetyloxyoxolan-2-yl methyl acetate, 2r,3s,4s,5s-5-acetoxymethyl tetrahydrofuran-2,3,4-triyl triacetate
+12.7° (c=2.4 in Chloroform)
[(2S,3R,4R,5S)-3,4,5-triacetyloxyoxolan-2-yl]methyl acetate
69431757
98%
1,2,3,5-Tetra-O-acetyl-beta-L-ribofuranose
80°C to 83°C
C13H18O9
1 g
IHNHAHWGVLXCCI-UMSGYPCISA-N
CC(=O)OCC1C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C
318.278
318.3
Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only