missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals 1,2,3,5-Tetra-O-acetyl-beta-L-ribofuranose, 98%
CAS: 144490-03-9 | C13H18O9 | 318.278 g/mol
Supplier: Thermo Scientific Chemicals J6607406
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1,2,3,5-Tetra-O-acetyl-beta-L-ribofuranose | |
| 80°C to 83°C | |
| C13H18O9 | |
| 5 g | |
| IHNHAHWGVLXCCI-UMSGYPCISA-N | |
| CC(=O)OCC1C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C | |
| 318.278 | |
| 318.3 | |
| Powder |
| 144490-03-9 | |
| White | |
| MFCD00674547 | |
| 1,2,3,5-tetra-o-acetyl-beta-l-ribofuranose, 1,2,3,5-tetra-o-acetyl-b-l-ribofuranose, 2s,3s,4s,5r-3,4,5-tris acetyloxy oxolan-2-yl methyl acetate, beta-l-ribofuranose 1,2,3,5-tetra-o-acetate, beta-l-ribofuranose tetraacetate, 1,2,3,5-tetra-o-acetyl-?-l-ribofuranose, 2s,3s,4s,5r-3,4,5-triacetyloxyoxolan-2-yl methyl acetate, 2r,3s,4s,5s-5-acetoxymethyl tetrahydrofuran-2,3,4-triyl triacetate | |
| +12.7° (c=2.4 in Chloroform) | |
| [(2S,3R,4R,5S)-3,4,5-triacetyloxyoxolan-2-yl]methyl acetate | |
| 69431757 | |
| 98% |
Chemical Identifiers
| 144490-03-9 | |
| 318.278 | |
| IHNHAHWGVLXCCI-UMSGYPCISA-N | |
| 69431757 | |
| CC(=O)OCC1C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C |
| C13H18O9 | |
| MFCD00674547 | |
| 1,2,3,5-tetra-o-acetyl-beta-l-ribofuranose, 1,2,3,5-tetra-o-acetyl-b-l-ribofuranose, 2s,3s,4s,5r-3,4,5-tris acetyloxy oxolan-2-yl methyl acetate, beta-l-ribofuranose 1,2,3,5-tetra-o-acetate, beta-l-ribofuranose tetraacetate, 1,2,3,5-tetra-o-acetyl-?-l-ribofuranose, 2s,3s,4s,5r-3,4,5-triacetyloxyoxolan-2-yl methyl acetate, 2r,3s,4s,5s-5-acetoxymethyl tetrahydrofuran-2,3,4-triyl triacetate | |
| [(2S,3R,4R,5S)-3,4,5-triacetyloxyoxolan-2-yl]methyl acetate |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only