missing translation for 'onlineSavingsMsg'
Learn More

1,2:3,5-Di-O-isopropylidene-alpha-D-xylofuranose 98.0+%, TCI America™

Catalog No. D26165G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D26165G Supplier TCI America Supplier No. D26165G
Only null left

Chemical Identifiers

CAS 20881-04-3
Molecular Formula C11H18O5
Molecular Weight (g/mol) 230.26
MDL Number MFCD00063224
InChI Key NKZDPBSWYPINNF-BZNPZCIMSA-N
Synonym 1,2:3,5-di-o-isopropylidene-alpha-d-xylofuranose, 1,2:3,5-di-o-isopropylidene-d-xylofuranose, 1,2:3,5-di-o-isopropylidene-a-d-xylofuranose, alpha-d-xylofuranose, 1,2:3,5-bis-o-1-methylethylidene, 1s,2r,6r,8r-4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo 6.4.0.0 2 ,? dodecane, xylofuranose, 1,2:3,5-di-o-isopropylidene-, .alpha.-d, .alpha.-d-xylofuranose, 1,2:3,5-bis-o-1-methylethylidene, 1,2,3,5-di-o-isopropylidene-alpha-d-xylofuranose, 1-o,2-o:3-o,5-o-diisopropylidene-alpha-d-xylofuranose, .alpha.-d-xylofuranose, 1,2,3,5-bis-o-1-methylethylidene
PubChem CID 30337
IUPAC Name (3aR,4aR,8aS,8bR)-2,2,7,7-tetramethyl-4a,5,8a,8b-tetrahydro-3aH-[1,3]dioxolo[3,4]furo[1,3-d][1,3]dioxine
SMILES CC1(OCC2C(O1)C3C(O2)OC(O3)(C)C)C

Specifications

Melting Point 45°C
Color White
Boiling Point 125°C
Quantity 5 g
Formula Weight 230.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2:3,5-Di-O-isopropylidene-alpha-D-xylofuranose