accessibility menu, dialog, popup

UserName

H3CCH3O(R)O(R)OCH3CH3O(R)(S)O

CAS RN 20881-04-3

IUPAC Name: (1S,2R,6R,8R)-4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo[6.4.0.0²,⁶]dodecane
Synonyms: (1S,2R,6R,8R)-4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo[6.4.0.0²,⁶]dodecane
Molecular Weight (g/mol): 230.26
Molecular Formula: C11H18O5
InChi Key: NKZDPBSWYPINNF-BZNPZCIMSA-N
SMILES: CC1(C)O[C@H]2O[C@@H]3COC(C)(C)O[C@@H]3[C@H]2O1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results