missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydroquinoxaline 98.0+%, TCI America™

Catalog No. T29995G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29995G 5 g
T29991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29995G Supplier TCI America Supplier No. T29995G

Chemical Identifiers

CAS 3476-89-9
Molecular Formula C8H10N2
Molecular Weight (g/mol) 134.18
MDL Number MFCD00047564
InChI Key HORKYAIEVBUXGM-UHFFFAOYSA-N
Synonym 1,2,3,4-tetrahydro-quinoxaline, tetrahydroquinoxaline, quinoxaline, 1,2,3,4-tetrahydro, 1,3,4-tetrahydroquinoxaline, ksc495i0b, 1,2,3,4-terahydroquinoxaline, quinoxaline,2,3,4-tetrahydro, 1,2,3,4-tetrahydro quinoxaline, benzo 2,3 piperazine, tetrahydro
PubChem CID 77028
IUPAC Name 1,2,3,4-tetrahydroquinoxaline
SMILES C1CNC2=CC=CC=C2N1

Specifications

Melting Point 97°C
Boiling Point 154°C
Quantity 5 g
Formula Weight 134.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3,4-Tetrahydroquinoxaline