accessibility menu, dialog, popup

UserName

HNNH

CAS RN 3476-89-9

IUPAC Name: 1,2,3,4-tetrahydroquinoxaline
Synonyms: 1,2,3,4-tetrahydroquinoxaline
Molecular Weight (g/mol): 134.18
Molecular Formula: C8H10N2
InChi Key: HORKYAIEVBUXGM-UHFFFAOYSA-N
SMILES: C1CNC2=CC=CC=C2N1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)