missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydroquinoxaline, 98%

Catalog No. p-7036972
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Catalog No. Quantity
AAH55473MD 250 mg
AAH5547303 1 g
AAH5547306 5 g
3 options

Catalog No. AAH55473MD

Supplier: Thermo Scientific Chemicals H55473MD

Only null left

CAS: 3476-89-9 | C8H10N2 | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3476-89-9
Molecular Formula C8H10N2
Molecular Weight (g/mol) 134.18
MDL Number MFCD00047564
InChI Key HORKYAIEVBUXGM-UHFFFAOYSA-N
Synonym 1,2,3,4-tetrahydro-quinoxaline, tetrahydroquinoxaline, quinoxaline, 1,2,3,4-tetrahydro, 1,3,4-tetrahydroquinoxaline, ksc495i0b, 1,2,3,4-terahydroquinoxaline, quinoxaline,2,3,4-tetrahydro, 1,2,3,4-tetrahydro quinoxaline, benzo 2,3 piperazine, tetrahydro
PubChem CID 77028
IUPAC Name 1,2,3,4-tetrahydroquinoxaline
SMILES C1CNC2=CC=CC=C2N1

Specifications

Melting Point 92°C to 98°C
Quantity 250 mg
Formula Weight 134.18
Percent Purity 98%
Chemical Name or Material 1,2,3,4-Tetrahydroquinoxaline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only