missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydroquinoline 95.0+%, TCI America™

Catalog No. p-7136089
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T011325ML 25 mL
T0113100ML 100 mL
T0113500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T011325ML Supplier TCI America Supplier No. T011325ML
Only null left

Chemical Identifiers

CAS 635-46-1
Molecular Formula C9H11N
Molecular Weight (g/mol) 133.19
MDL Number MFCD00006693
InChI Key LBUJPTNKIBCYBY-UHFFFAOYSA-N
Synonym tetrahydroquinoline, 1,2,3,4-tetrahydro-quinoline, kusol, quinoline, 1,2,3,4-tetrahydro, unii-ccr50n1z9g, 1,2,3,4-tetrahydrochinoline, 1,2,3,4-tetrahydro quinoline, benzopiperidine, ccr50n1z9g, 1,2,3,4,-tetrahydroquinoline
PubChem CID 69460
ChEBI CHEBI:213323
IUPAC Name 1,2,3,4-tetrahydroquinoline
SMILES C1CNC2=CC=CC=C2C1

Specifications

Color Yellow
Boiling Point 251°C
Quantity 25 mL
Formula Weight 133.19
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2,3,4-Tetrahydroquinoline
TSCA Yes
Recommended Storage Refrigerator