accessibility menu, dialog, popup

UserName

HN

CAS RN 635-46-1

IUPAC Name: 1,2,3,4-tetrahydroquinoline
Synonyms: tetrahydroquinoline
Molecular Weight (g/mol): 133.19
Molecular Formula: C9H11N
InChi Key: LBUJPTNKIBCYBY-UHFFFAOYSA-N
SMILES: C1CNC2=CC=CC=C2C1
Boiling Point 
  • (2)
  • (4)
  • (3)
Percent Purity 
  • (3)
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (2)
  • (4)
  • (3)
  • (3)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (7)