missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3,4-Tetrahydrocarbazole 99.0+%, TCI America™
$318.75 - $959.98
Chemical Identifiers
| CAS | 942-01-8 |
|---|---|
| Molecular Formula | C12H13N |
| Molecular Weight (g/mol) | 171.243 |
| MDL Number | MFCD00004959 |
| InChI Key | XKLNOVWDVMWTOB-UHFFFAOYSA-N |
| Synonym | 1,2,3,4-tetrahydrocarbazole, 2,3-tetramethyleneindole, 2,3-tetramethylene-1h-indole, 5,6,7,8-tetrahydrocarbazole, tetrahydrocarbazole, 1h-carbazole, 2,3,4,9-tetrahydro, carbazole, 1,2,3,4-tetrahydro, 1h-indole, 2,3-1,4-butanediyl, carbazole, 5,6,7,8-tetrahydro, unii-8zlk0tsx93 |
| PubChem CID | 13664 |
| IUPAC Name | 2,3,4,9-tetrahydro-1H-carbazole |
| SMILES | C1CCC2=C(C1)C3=CC=CC=C3N2 |
Chemical Identifiers
| 942-01-8 | |
| 171.243 | |
| XKLNOVWDVMWTOB-UHFFFAOYSA-N | |
| 13664 | |
| C1CCC2=C(C1)C3=CC=CC=C3N2 |
| C12H13N | |
| MFCD00004959 | |
| 1,2,3,4-tetrahydrocarbazole, 2,3-tetramethyleneindole, 2,3-tetramethylene-1h-indole, 5,6,7,8-tetrahydrocarbazole, tetrahydrocarbazole, 1h-carbazole, 2,3,4,9-tetrahydro, carbazole, 1,2,3,4-tetrahydro, 1h-indole, 2,3-1,4-butanediyl, carbazole, 5,6,7,8-tetrahydro, unii-8zlk0tsx93 | |
| 2,3,4,9-tetrahydro-1H-carbazole |
Specifications
| 942-01-8 | |
| White-Yellow | |
| C12H13N | |
| 25 g | |
| XKLNOVWDVMWTOB-UHFFFAOYSA-N | |
| 2,3,4,9-tetrahydro-1H-carbazole | |
| 13664 | |
| ≥99.0% (GC) | |
| 1,2,3,4-Tetrahydrocarbazole |
| 119°C | |
| 186°C | |
| MFCD00004959 | |
| 1,2,3,4-tetrahydrocarbazole, 2,3-tetramethyleneindole, 2,3-tetramethylene-1h-indole, 5,6,7,8-tetrahydrocarbazole, tetrahydrocarbazole, 1h-carbazole, 2,3,4,9-tetrahydro, carbazole, 1,2,3,4-tetrahydro, 1h-indole, 2,3-1,4-butanediyl, carbazole, 5,6,7,8-tetrahydro, unii-8zlk0tsx93 | |
| C1CCC2=C(C1)C3=CC=CC=C3N2 | |
| 171.243 | |
| 171.24 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : FE6300000
TSCA : No