missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3,4-Tetrahydrocarbazol-4-one 98.0+%, TCI America™
$31.68 - $91.33
Chemical Identifiers
| CAS | 15128-52-6 |
|---|---|
| Molecular Formula | C12H11NO |
| Molecular Weight (g/mol) | 185.226 |
| MDL Number | MFCD00173749 |
| InChI Key | DSXKDTZEIWTHRO-UHFFFAOYSA-N |
| Synonym | 1,2,3,4-Tetrahydro-4-oxocarbazole |
| PubChem CID | 929130 |
| IUPAC Name | 1,2,3,9-tetrahydrocarbazol-4-one |
| SMILES | C1CC2=C(C(=O)C1)C3=CC=CC=C3N2 |
Chemical Identifiers
| 15128-52-6 | |
| 185.226 | |
| DSXKDTZEIWTHRO-UHFFFAOYSA-N | |
| 929130 | |
| C1CC2=C(C(=O)C1)C3=CC=CC=C3N2 |
| C12H11NO | |
| MFCD00173749 | |
| 1,2,3,4-Tetrahydro-4-oxocarbazole | |
| 1,2,3,9-tetrahydrocarbazol-4-one |
Specifications
| 15128-52-6 | |
| C12H11NO | |
| 5 g | |
| DSXKDTZEIWTHRO-UHFFFAOYSA-N | |
| 1,2,3,9-tetrahydrocarbazol-4-one | |
| 929130 | |
| ≥98.0% (GC) | |
| 1,2,3,4-Tetrahydrocarbazol-4-one |
| 226°C | |
| MFCD00173749 | |
| 1,2,3,4-Tetrahydro-4-oxocarbazole | |
| C1CC2=C(C(=O)C1)C3=CC=CC=C3N2 | |
| 185.226 | |
| 185.23 | |
| Crystalline Powder |
Safety and Handling
TSCA : No