accessibility menu, dialog, popup

UserName

OHN

CAS RN 15128-52-6

IUPAC Name: 2,3,4,9-tetrahydro-1H-carbazol-4-one
Synonyms: 1,2,3,9-tetrahydrocarbazol-4-one
Molecular Weight (g/mol): 185.23
Molecular Formula: C12H11NO
InChi Key: DSXKDTZEIWTHRO-UHFFFAOYSA-N
SMILES: O=C1CCCC2=C1C1=CC=CC=C1N2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results