missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydrocarbazol-4-one 98.0+%, TCI America™

Catalog No. T25595G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25595G 5 g
T255925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25595G Supplier TCI America Supplier No. T25595G
Only null left

Chemical Identifiers

CAS 15128-52-6
Molecular Formula C12H11NO
Molecular Weight (g/mol) 185.226
MDL Number MFCD00173749
InChI Key DSXKDTZEIWTHRO-UHFFFAOYSA-N
Synonym 1,2,3,4-Tetrahydro-4-oxocarbazole
PubChem CID 929130
IUPAC Name 1,2,3,9-tetrahydrocarbazol-4-one
SMILES C1CC2=C(C(=O)C1)C3=CC=CC=C3N2

Specifications

Melting Point 226°C
Quantity 5 g
Formula Weight 185.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3,4-Tetrahydrocarbazol-4-one