missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydro-2-naphthylamine, 97%, Thermo Scientific Chemicals

Catalog No. AAH6665203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6665203 1 g
AAH6665206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6665203 Supplier Thermo Scientific Chemicals Supplier No. H6665203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2954-50-9 | Molecular Formula: C10H13N | Molecular Weight: 147.221 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2954-50-9
Molecular Formula C10H13N
Molecular Weight (g/mol) 147.221
MDL Number MFCD00045577
InChI Key LCGFVWKNXLRFIF-UHFFFAOYSA-N
Synonym 1,2,3,4-tetrahydro-2-naphthylamine, 2-aminotetralin, 2-naphthalenamine, 1,2,3,4-tetrahydro, 1,2,3,4-tetrahydro-2-naphthalenamine, 1,2,3,4-tetrahydro-naphthalen-2-ylamine, chembl30294, 2-1,2,3,4-tetrahydronaphthylamine, 2-amino-1,2,3,4-tetrahydronaphthalene, 2-amino-tetralin, tetralin-2-amine
PubChem CID 34677
IUPAC Name 1,2,3,4-tetrahydronaphthalen-2-amine
SMILES C1CC2=CC=CC=C2CC1N

Specifications

Quantity 1 g
Beilstein 1102357
Formula Weight 147.1
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1,2,3,4-Tetrahydro-2-naphthylamine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 220-973-7
RTECSNumber QM4900000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only