missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3,4-Tetrahydro-1-naphthylamine 98.0+%, TCI America™
Supplier: TCI America T199225G
Specifications
| 1,2,3,4-Tetrahydro-1-naphthylamine | |
| Yellow | |
| C10H14ClN | |
| 25 g | |
| DETWFIUAXSWCIK-UHFFFAOYNA-N | |
| 1,2,3,4-tetrahydronaphthalen-1-aminium chloride | |
| 18066 | |
| ≥98.0% (GC,T) |
| 2217-40-5 | |
| 230°C | |
| MFCD00001740 | |
| 1,2,3,4-tetrahydro-1-naphthylamine, 1-aminotetralin, aminotetralin, 1-amino-1,2,3,4-tetrahydro-naphthalene, aminotetralin czech, tetrahydro-beta-naphthylamine, 1-amino-1,2,3,4-tetrahydronaphthalene, 1-naphthalenamine, 1,2,3,4-tetrahydro, 1,2,3,4-tetrahydronaphthylamine, beta-1,2,3,4-tetrahydronaphthylamine | |
| [Cl-].[NH3+]C1CCCC2=CC=CC=C12 | |
| 183.68 | |
| 147.22 | |
| Liquid |
Chemical Identifiers
| 2217-40-5 | |
| 183.68 | |
| DETWFIUAXSWCIK-UHFFFAOYNA-N | |
| 18066 | |
| [Cl-].[NH3+]C1CCCC2=CC=CC=C12 |
| C10H14ClN | |
| MFCD00001740 | |
| 1,2,3,4-tetrahydro-1-naphthylamine, 1-aminotetralin, aminotetralin, 1-amino-1,2,3,4-tetrahydro-naphthalene, aminotetralin czech, tetrahydro-beta-naphthylamine, 1-amino-1,2,3,4-tetrahydronaphthalene, 1-naphthalenamine, 1,2,3,4-tetrahydro, 1,2,3,4-tetrahydronaphthylamine, beta-1,2,3,4-tetrahydronaphthylamine | |
| 1,2,3,4-tetrahydronaphthalen-1-aminium chloride |
Safety and Handling
TSCA : No