missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1,2,3,4-Tetrahydro-1-naphthylamine, 97%

Catalog No. AAB2567306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2567306 5 g
AAB2567314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2567306 Supplier Thermo Scientific Chemicals Supplier No. B2567306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2217-40-5 | C10H14ClN | 183.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2217-40-5
Molecular Formula C10H14ClN
Molecular Weight (g/mol) 183.68
MDL Number MFCD00001740
InChI Key DETWFIUAXSWCIK-UHFFFAOYNA-N
Synonym 1,2,3,4-tetrahydro-1-naphthylamine, 1-aminotetralin, aminotetralin, 1-amino-1,2,3,4-tetrahydro-naphthalene, aminotetralin czech, tetrahydro-beta-naphthylamine, 1-amino-1,2,3,4-tetrahydronaphthalene, 1-naphthalenamine, 1,2,3,4-tetrahydro, 1,2,3,4-tetrahydronaphthylamine, beta-1,2,3,4-tetrahydronaphthylamine
PubChem CID 18066
IUPAC Name 1,2,3,4-tetrahydronaphthalen-1-amine
SMILES [Cl-].[NH3+]C1CCCC2=CC=CC=C12

Specifications

Density 1.026
Boiling Point 246°C to 247°C (714 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.564
Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 147.22
Percent Purity 97%
Chemical Name or Material (+/-)-1,2,3,4-Tetrahydro-1-naphthylamine
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 218-712-7
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only