missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydro-1-naphthol 95.0+%, TCI America™

Catalog No. p-7136614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11115G 5 g
T111125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T11115G Supplier TCI America Supplier No. T11115G

Chemical Identifiers

CAS 529-33-9
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.21
MDL Number MFCD00001739,MFCD00063006
InChI Key JAAJQSRLGAYGKZ-UHFFFAOYNA-N
Synonym 1,2,3,4-tetrahydro-1-naphthol, tetralin-1-ol, 1-tetralol, alpha-tetralol, 1-hydroxytetralin, 1-naphthalenol, 1,2,3,4-tetrahydro, .alpha.-tetralol, tetrahydro-1-naphthol van, +-1,2,3,4-tetrahydro-1-naphthol, tetrahydro-1-naphthol
PubChem CID 10723
IUPAC Name 1,2,3,4-tetrahydronaphthalen-1-ol
SMILES OC1CCCC2=CC=CC=C12

Specifications

Color White-Yellow
Boiling Point 104°C
Quantity 5 g
Formula Weight 148.21
Percent Purity ≥95.0% (GC)
Physical Form Lumps
Chemical Name or Material 1,2,3,4-Tetrahydro-1-naphthol
RTECSNumber QL5075000
TSCA Yes