missing translation for 'onlineSavingsMsg'
Learn More

1,2:3,4-Di-O-isopropylidene-alpha-D-galactopyranose 92.0+%, TCI America™

Catalog No. p-7127365
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25555G 5 g
D255525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25555G Supplier TCI America Supplier No. D25555G
Only null left

Chemical Identifiers

CAS 4064-06-6
Molecular Formula C12H20O6
Molecular Weight (g/mol) 260.29
MDL Number MFCD00063225
InChI Key POORJMIIHXHXAV-UHFFFAOYNA-N
Synonym 1,2:3,4-di-o-isopropylidene-d-galactose, 1,2:3,4-di-o-isopropylidene-, a-d-galactopyranose, 1,2:3,4-bis-o-1-methylethylidene-, 1s,6r,9s-4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo 7.3.0.0 2 ,? dodecan-8-yl methanol
PubChem CID 70793572
IUPAC Name [(3aR,5aS,8aS)-2,2,7,7-tetramethyl-5,5a,8a,8b-tetrahydro-3aH-di[1,3]dioxolo[4,5-a
SMILES CC1(OC2C(OC3C(C2O1)OC(O3)(C)C)CO)C

Specifications

Color Yellow
Boiling Point 133°C
Quantity 5 g
Formula Weight 260.29
Percent Purity ≥92.0% (GC)
Physical Form Starch Syrup
Chemical Name or Material 1,2:3,4-Di-O-isopropylidene-alpha-D-galactopyranose