missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3,4-Cyclopentanetetracarboxylic Dianhydride 98.0+%, TCI America™
$240.46 - $959.98
Chemical Identifiers
| CAS | 6053-68-5 |
|---|---|
| Molecular Formula | C9H6O6 |
| Molecular Weight (g/mol) | 210.141 |
| MDL Number | MFCD00046872 |
| InChI Key | NLWBEORDOPDUPM-UHFFFAOYSA-N |
| PubChem CID | 110757 |
| SMILES | C1C2C(C3C1C(=O)OC3=O)C(=O)OC2=O |
Chemical Identifiers
| 6053-68-5 | |
| 210.141 | |
| NLWBEORDOPDUPM-UHFFFAOYSA-N | |
| C1C2C(C3C1C(=O)OC3=O)C(=O)OC2=O |
| C9H6O6 | |
| MFCD00046872 | |
| 110757 |
Specifications
| 6053-68-5 | |
| White | |
| MFCD00046872 | |
| NLWBEORDOPDUPM-UHFFFAOYSA-N | |
| 210.141 | |
| 210.14 | |
| Crystalline Powder |
| 213°C | |
| C9H6O6 | |
| 5 g | |
| C1C2C(C3C1C(=O)OC3=O)C(=O)OC2=O | |
| 110757 | |
| ≥98.0% (T) | |
| 1,2,3,4-Cyclopentanetetracarboxylic Dianhydride |
Safety and Handling
TSCA : Yes