missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3,4-Cyclopentanetetracarboxylic Dianhydride 98.0+%, TCI America™
Supplier: TCI America C085725G
Specifications
| 1,2,3,4-Cyclopentanetetracarboxylic Dianhydride | |
| 213°C | |
| C9H6O6 | |
| 25 g | |
| C1C2C(C3C1C(=O)OC3=O)C(=O)OC2=O | |
| 110757 | |
| ≥98.0% (T) |
| 6053-68-5 | |
| White | |
| MFCD00046872 | |
| NLWBEORDOPDUPM-UHFFFAOYSA-N | |
| 210.141 | |
| 210.14 | |
| Crystalline Powder |
Chemical Identifiers
| 6053-68-5 | |
| 210.141 | |
| NLWBEORDOPDUPM-UHFFFAOYSA-N | |
| C1C2C(C3C1C(=O)OC3=O)C(=O)OC2=O |
| C9H6O6 | |
| MFCD00046872 | |
| 110757 |
Safety and Handling
TSCA : Yes