missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Cyclopentanetetracarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7124646
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C08565G 5 g
C085625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C08565G Supplier TCI America Supplier No. C08565G

Chemical Identifiers

CAS 3724-52-5
Molecular Formula C9H10O8
Molecular Weight (g/mol) 246.171
MDL Number MFCD00001377
InChI Key WOSVXXBNNCUXMT-UHFFFAOYSA-N
PubChem CID 19622
IUPAC Name cyclopentane-1,2,3,4-tetracarboxylic acid
SMILES C1C(C(C(C1C(=O)O)C(=O)O)C(=O)O)C(=O)O

Specifications

Color White
Quantity 5 g
Formula Weight 246.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3,4-Cyclopentanetetracarboxylic Acid
RTECSNumber GY4700500
TSCA No