accessibility menu, dialog, popup

UserName

OHOHOOOHOHOO

CAS RN 3724-52-5

IUPAC Name: cyclopentane-1,2,3,4-tetracarboxylic acid
Synonyms: cyclopentane-1,2,3,4-tetracarboxylic acid
Molecular Weight (g/mol): 246.17
Molecular Formula: C9H10O8
InChi Key: WOSVXXBNNCUXMT-UHFFFAOYNA-N
SMILES: OC(=O)C1CC(C(C1C(O)=O)C(O)=O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results