missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Cyclobutanetetracarboxylic Dianhydride (purified by sublimation) 98.0+%, TCI America™

Catalog No. p-7125679
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28421G 1 g
C28425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28421G Supplier TCI America Supplier No. C28421G
Only null left

Chemical Identifiers

CAS 4415-87-6
Molecular Formula C8H4O6
Molecular Weight (g/mol) 196.114
MDL Number MFCD00004944
InChI Key YGYCECQIOXZODZ-UHFFFAOYSA-N
PubChem CID 107280
SMILES C12C(C3C1C(=O)OC3=O)C(=O)OC2=O

Specifications

Melting Point 285°C
Color White-Yellow
Quantity 1 g
Formula Weight 196.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3,4-Cyclobutanetetracarboxylic Dianhydride (purified by sublimation)
EINECSNumber (5)-6721
TSCA No