missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3,4,5,6-Cyclohexanehexacarboxylic Acid Monohydrate 97.0+%, TCI America™
Supplier: TCI America C142725G
Specifications
| 1,2,3,4,5,6-Cyclohexanehexacarboxylic Acid Monohydrate | |
| White | |
| MFCD00081090 | |
| DTGRIEIJTWNZQF-UHFFFAOYSA-N | |
| cyclohexane-1,2,3,4,5,6-hexacarboxylic acid | |
| 102227 | |
| ≥97.0% (T) |
| 2216-84-4 | |
| C12H12O12 | |
| 25 g | |
| C1(C(C(C(C(C1C(=O)O)C(=O)O)C(=O)O)C(=O)O)C(=O)O)C(=O)O | |
| 348.216 | |
| 348.22 | |
| Crystalline Powder |
Chemical Identifiers
| 2216-84-4 | |
| 348.216 | |
| DTGRIEIJTWNZQF-UHFFFAOYSA-N | |
| cyclohexane-1,2,3,4,5,6-hexacarboxylic acid |
| C12H12O12 | |
| MFCD00081090 | |
| 102227 | |
| C1(C(C(C(C(C1C(=O)O)C(=O)O)C(=O)O)C(=O)O)C(=O)O)C(=O)O |
Safety and Handling
RTECSNumber : GV3100000
TSCA : No