missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4,5,6-Cyclohexanehexacarboxylic Acid Monohydrate 97.0+%, TCI America™

Catalog No. p-7124972
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14275G 5 g
C142725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C14275G Supplier TCI America Supplier No. C14275G
Only null left

Chemical Identifiers

CAS 2216-84-4
Molecular Formula C12H12O12
Molecular Weight (g/mol) 348.216
MDL Number MFCD00081090
InChI Key DTGRIEIJTWNZQF-UHFFFAOYSA-N
PubChem CID 102227
IUPAC Name cyclohexane-1,2,3,4,5,6-hexacarboxylic acid
SMILES C1(C(C(C(C(C1C(=O)O)C(=O)O)C(=O)O)C(=O)O)C(=O)O)C(=O)O

Specifications

Color White
Quantity 5 g
Formula Weight 348.22
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3,4,5,6-Cyclohexanehexacarboxylic Acid Monohydrate
RTECSNumber GV3100000
TSCA No