missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,10b-Tetrahydrofluoranthene 98.0+%, TCI America™

Catalog No. T07711G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T07711G 1 g
T07715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T07711G Supplier TCI America Supplier No. T07711G
Only null left

Chemical Identifiers

CAS 20279-21-4
Molecular Formula C16H14
Molecular Weight (g/mol) 206.29
MDL Number MFCD00045348
InChI Key VMBZUGDYWLFLEU-UHFFFAOYNA-N
PubChem CID 89305
IUPAC Name 1,2,3,10b-tetrahydrofluoranthene
SMILES C1CC2C3=CC=CC=C3C3=CC=CC(C1)=C23

Specifications

Melting Point 76°C
Color White-Yellow
Boiling Point 205°C
Quantity 1 g
Formula Weight 206.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3,10b-Tetrahydrofluoranthene