missing translation for 'onlineSavingsMsg'
Learn More

1,2,2,3-Propanetetracarbonitrile 96.0+%, TCI America™

Catalog No. P17511G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17511G 1 g
P17515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P17511G Supplier TCI America Supplier No. P17511G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1274904-48-1
Molecular Formula C7H4N4
Molecular Weight (g/mol) 144.14
MDL Number MFCD10000963
InChI Key RAFBXJGDOLMWDJ-UHFFFAOYSA-N
Synonym 1,2,2,3-Tetracyanopropane
PubChem CID 44629878
IUPAC Name propane-1,2,2,3-tetracarbonitrile
SMILES N#CCC(CC#N)(C#N)C#N

Specifications

Quantity 1 g
UN Number 3439
Formula Weight 144.14
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,2,3-Propanetetracarbonitrile