accessibility menu, dialog, popup

UserName

NNNN

CAS RN 1274904-48-1

IUPAC Name: propane-1,2,2,3-tetracarbonitrile
Synonyms: propane-1,2,2,3-tetracarbonitrile
Molecular Weight (g/mol): 144.14
Molecular Formula: C7H4N4
InChi Key: RAFBXJGDOLMWDJ-UHFFFAOYSA-N
SMILES: N#CCC(CC#N)(C#N)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results