missing translation for 'onlineSavingsMsg'
Learn More

1,13-Tetradecadiene 90.0+%, TCI America™

Catalog No. T06731G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T06731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T06731G Supplier TCI America Supplier No. T06731G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 21964-49-8
Molecular Formula C14H26
Molecular Weight (g/mol) 194.362
MDL Number MFCD00014941
InChI Key XMRSTLBCBDIKFI-UHFFFAOYSA-N
Synonym 1,13-tetradecadiene, tetradecadiene-1,13, acmc-209fpz
PubChem CID 30875
IUPAC Name tetradeca-1,13-diene
SMILES C=CCCCCCCCCCCC=C

Specifications

Color Yellow
Boiling Point 131°C
Quantity 1 g
Formula Weight 194.36
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,13-Tetradecadiene
EINECSNumber (2)-3621
TSCA No