CAS RN 21964-49-8
IUPAC Name:
tetradeca-1,13-diene
Synonyms:
tetradeca-1,13-diene
Molecular Weight (g/mol):
194.36
Molecular Formula:
C14H26
InChi Key:
XMRSTLBCBDIKFI-UHFFFAOYSA-N
SMILES:
C=CCCCCCCCCCCC=C
1
–
2
of
2
results
1
Promotions Available
1,13-Tetradecadiene, 90+%
1,13-Tetradecadiene, 90+%
CAS: 21964-49-8 Molecular Formula: C14H26 Molecular Weight (g/mol): 194.362 MDL Number: MFCD00014941 InChI Key: XMRSTLBCBDIKFI-UHFFFAOYSA-N Synonym: 1,13-tetradecadiene,tetradecadiene-1,13,acmc-209fpz PubChem CID: 30875 IUPAC Name: tetradeca-1,13-diene SMILES: C=CCCCCCCCCCCC=C
2
1,13-Tetradecadiene 90.0+%, TCI America™
CAS: 21964-49-8 Molecular Formula: C14H26 Molecular Weight (g/mol): 194.362 MDL Number: MFCD00014941 InChI Key: XMRSTLBCBDIKFI-UHFFFAOYSA-N Synonym: 1,13-tetradecadiene,tetradecadiene-1,13,acmc-209fpz PubChem CID: 30875 IUPAC Name: tetradeca-1,13-diene SMILES: C=CCCCCCCCCCCC=C