missing translation for 'onlineSavingsMsg'
Learn More

1,12-Diacetoxydodecane 98.0+%, TCI America™

Catalog No. D33045G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D33045G Supplier TCI America Supplier No. D33045G
Only null left

Chemical Identifiers

CAS 42236-50-0
Molecular Formula C16H30O4
Molecular Weight (g/mol) 286.412
InChI Key VSQSFRDZIRKBEF-UHFFFAOYSA-N
Synonym Dodecamethylene Diacetate, 1,12-Dodecanediol Diacetate
PubChem CID 20788573
IUPAC Name 12-acetyloxydodecyl acetate
SMILES CC(=O)OCCCCCCCCCCCCOC(=O)C

Specifications

Melting Point 40°C
Color White
Boiling Point 190°C
Quantity 5 g
Formula Weight 286.41
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,12-Diacetoxydodecane
TSCA No
Recommended Storage Refrigerator