accessibility menu, dialog, popup

UserName

CAS RN 42236-50-0

H3COOOOCH3

CAS RN 42236-50-0

IUPAC Name: 12-(acetyloxy)dodecyl acetate
Synonyms: 12-(acetyloxy)dodecyl acetate
Molecular Weight (g/mol): 286.41
Molecular Formula: C16H30O4
InChi Key: VSQSFRDZIRKBEF-UHFFFAOYSA-N
SMILES: CC(=O)OCCCCCCCCCCCCOC(C)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results