missing translation for 'onlineSavingsMsg'
Learn More

1,11-Undecanedicarboxylic acid, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 369440050

 View more versions of this product

Catalog No. AC369440050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

1, 11-Undecanedicarboxylic acid
505-52-2
100.0
94%
C13H24O4
MFCD00002740
02,731
Solubility in water: insoluble. Other solubilities: soluble in ethanol,ether
C(CCCCCC(=O)O)CCCCCC(=O)O
244.33
CHEBI:73718
≥92%
94%
92.0
Authentic
Glass bottle
HO2C(CH2)11CO2H
5g
brassylic acid, 1,11-undecanedicarboxylic acid, brassilic acid, 1,13-tridecanedioic acid, brassylate, unii-pl3iq40c34, undecane-1,11-dicarboxylic acid, tridecanedioate, dsstox_cid_1683, dsstox_rid_76281
DXNCZXXFRKPEPY-UHFFFAOYSA-N
tridecanedioic acid
10458
244.33

Chemical Identifiers

505-52-2
244.33
DXNCZXXFRKPEPY-UHFFFAOYSA-N
10458
tridecanedioic acid
C13H24O4
MFCD00002740
brassylic acid, 1,11-undecanedicarboxylic acid, brassilic acid, 1,13-tridecanedioic acid, brassylate, unii-pl3iq40c34, undecane-1,11-dicarboxylic acid, tridecanedioate, dsstox_cid_1683, dsstox_rid_76281
CHEBI:73718
C(CCCCCC(=O)O)CCCCCC(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/pro

GHS Signal Word: Warning

EINECSNumber : 208-011-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only