missing translation for 'onlineSavingsMsg'
Learn More

1,11-Dioxa[11]paracyclophane 97.0+%, TCI America™

Catalog No. p-7128207
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4298250MG 250 mg
D42981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4298250MG Supplier TCI America Supplier No. D4298250MG

Chemical Identifiers

CAS 6571-51-3
Molecular Formula C15H22O2
Molecular Weight (g/mol) 234.34
MDL Number MFCD28100826
InChI Key LTROLYACVISZRU-UHFFFAOYSA-N
PubChem CID 91972135
IUPAC Name 2,12-dioxabicyclo[11.2.2]heptadeca-1(15),13,16-triene
SMILES C1CCCCOC2=CC=C(OCCCC1)C=C2

Specifications

Melting Point 55°C
Color White-Yellow
Quantity 250 mg
Formula Weight 234.34
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,11-Dioxa[11]paracyclophane