accessibility menu, dialog, popup

UserName

OO

CAS RN 6571-51-3

IUPAC Name: 2,12-dioxabicyclo[11.2.2]heptadeca-1(15),13,16-triene
Synonyms: 2,12-dioxabicyclo[11.2.2]heptadeca-1(15),13,16-triene
Molecular Weight (g/mol): 234.34
Molecular Formula: C15H22O2
InChi Key: LTROLYACVISZRU-UHFFFAOYSA-N
SMILES: C1CCCCOC2=CC=C(OCCCC1)C=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results