missing translation for 'onlineSavingsMsg'
Learn More

(1,10-Phenanthroline)tris[4,4,4-trifluoro-1-(2-thienyl)-1,3-butanedionato]europium(III) 98.0+%, TCI America™

Catalog No. P17661G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P17661G Supplier TCI America Supplier No. P17661G
Only null left

Chemical Identifiers

CAS 17904-86-8
Molecular Formula C36H20EuF9N2O6S3
Molecular Weight (g/mol) 995.694
InChI Key GMFTYFSOONOZOH-MCTJRNESSA-K
Synonym Eu(TTA)3phen
PubChem CID 15369070
IUPAC Name europium(3+);1,10-phenanthroline;(Z)-4,4,4-trifluoro-3-oxo-1-thiophen-2-ylbut-1-en-1-olate
SMILES C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.C1=CSC(=C1)C(=CC(=O)C(F)(F)F)[O-].C1=CSC(=C1)C(=CC(=O)C(F)(F)F)[O-].C1=CSC(=C1)C(=CC(=O)C(F)(F)F)[O-].[Eu+3]

Specifications

Melting Point 248°C
Quantity 1 g
Formula Weight 995.69
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (1,10-Phenanthroline)tris[4,4,4-trifluoro-1-(2-thienyl)-1,3-butanedionato]europium(III)