CAS RN 17904-86-8
CAS RN 17904-86-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(1,10-Phenanthroline)tris[4,4,4-trifluoro-1-(2-thienyl)-1,3-butanedionato]europium(III) 98.0+%, TCI America™
CAS: 17904-86-8 Molecular Formula: C36H20EuF9N2O6S3 Molecular Weight (g/mol): 995.694 InChI Key: GMFTYFSOONOZOH-MCTJRNESSA-K Synonym: Eu(TTA)3phen PubChem CID: 15369070 IUPAC Name: europium(3+);1,10-phenanthroline;(Z)-4,4,4-trifluoro-3-oxo-1-thiophen-2-ylbut-1-en-1-olate SMILES: C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.C1=CSC(=C1)C(=CC(=O)C(F)(F)F)[O-].C1=CSC(=C1)C(=CC(=O)C(F)(F)F)[O-].C1=CSC(=C1)C(=CC(=O)C(F)(F)F)[O-].[Eu+3]