missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline monohydrate, 99+%, pure

Catalog No. p-3737172
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC130130050 5 g Glass bottle
AC130130250 25 g Glass bottle
AC130131000 100 g Glass bottle
AC130135000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC130130050 Supplier Thermo Scientific Chemicals Supplier No. 130150050
Only null left

CAS: 5144-89-8 | C12H10N2O | 198.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 5144-89-8 , 66-71-7
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.23
MDL Number MFCD00149973
InChI Key PPQJCISYYXZCAE-UHFFFAOYSA-N
PubChem CID 21226
SMILES O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 97.0°C to 101.0°C
Color Cream-White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 5 g
Beilstein 23,227
Merck Index 15,7328
Synonym 1,10-phenanthroline hydrate, 1,10-phenanthroline monohydrate, o-phenanthroline monohydrate, unii-ksx215x00e, 1,10-fenantrolina, 4,5-phenanthroline monohydrate, 1,10-phenanthroline, monohydrate, 1,10-fenanthrolin, 1,10-phenanthrolineo-phenanthroline, 1, 10-phenanthroline monohydrate
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, acetone and benzene
IUPAC Name 1,10-phenanthroline;hydrate
Formula Weight 198.23
Percent Purity 99+%
Grade Pure
Physical Form Needle-like Crystalline Powder
Chemical Name or Material 1, 10-Phenanthroline monohydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

RUO – Research Use Only