Learn More
1,10-Phenanthroline, 99%, may contain up to 1.5% water
CAS: 66-71-7 | C12H8N2 | 180.21 g/mol
Supplier: Thermo Scientific Chemicals A1316322
Description
1,10-Phenanthroline, anhydrous is used as a chelating agent, used as bidendate nitrogen ligands in Heck type reactions as an alternative to phospine ligands. It is widely used as an indicator for alkyllithium reagents and an inhibitor of metallopeptidase enzymes. It is also used in the analysis formulations in end-user industries like food, beverage and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,10-Phenanthroline is used as a chelating agent, used as bidendate nitrogen ligands in Heck type reactions as an alternative to phospine ligands. It is widely used as an indicator for alkyllithium reagents and an inhibitor of metallopeptidase enzymes. It is also used in the analysis formulations in end-user industries like food, beverage and pharmaceuticals.
Solubility
Soluble in acetone and ethanol. Slightly soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids. Store in a tightly closed container. Store in a cool, dry, well-ventilated area.
Specifications
| 1,10-Phenanthroline | |
| 116°C to 119°C | |
| Odorless | |
| MFCD00011678 | |
| UN2811 | |
| Hygroscopic | |
| o-phenanthroline, 4,5-diazaphenanthrene, orthophenanthroline, phenanthroline, 1,10-o-phenanthroline, phen, 2-phenanthroline, beta-phenanthroline, .beta.-phenanthroline | |
| DGEZNRSVGBDHLK-UHFFFAOYSA-N | |
| 1,10-phenanthroline | |
| 1318 | |
| 180.21 |
| 66-71-7 | |
| >330°C | |
| C12H8N2 | |
| 100 g | |
| 126461 | |
| 14,7214 | |
| Soluble in acetone and ethanol. Slightly soluble in water. | |
| C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1 | |
| 180.21 | |
| CHEBI:44975 | |
| 99% |
Chemical Identifiers
| 66-71-7 | |
| 180.21 | |
| DGEZNRSVGBDHLK-UHFFFAOYSA-N | |
| 1318 | |
| 1,10-phenanthroline |
| C12H8N2 | |
| MFCD00011678 | |
| o-phenanthroline, 4,5-diazaphenanthrene, orthophenanthroline, phenanthroline, 1,10-o-phenanthroline, phen, 2-phenanthroline, beta-phenanthroline, .beta.-phenanthroline | |
| CHEBI:44975 | |
| C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1 |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P310-P330-P501c
H301-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 200-629-2
RTECSNumber : SF8300000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only